Environmental Express Katanax® Platinumware Polishing Kit; 230 VAC, 50 Hz

Representative image only.

Environmental Express Katanax® Platinumware Polishing Kit; 230 VAC, 50 Hz

Environmental Express | Mfr: KP9005A| Item: 0457879

Polish the surfaces of your crucibles and molds to maintain high standards in your sample preparation process

$2,232.19CAD / Each

$2,232.19CAD / Each
Product Description

Includes

Electric handheld rotary tool, flexible shaft, table stand, sets of buffing pads, and diamond pastes of assorted grades.

Key Features

  • Restore the shiny finish to platinumware of any shape and size
  • 230 VAC, 50 Hz operation
  • 1 year warranty

More About this Item

Proper maintenance of your platinumware will extend the service life as well as ensure great results.

1 Year Warranty
Specifications
  • Compatible WithAll types of platinumware
  • Power (VAC)230
  • Power (Hz)50
  • DescriptionPlatinumware Polishing Kit; 230 VAC, 50 Hz
  • Warranty1 Year
Reviews
Q&A