Environmental Express Katanax® Platinumware Polishing Kit; 115 VAC, 60 Hz

Representative image only.

Environmental Express Katanax® Platinumware Polishing Kit; 115 VAC, 60 Hz

Environmental Express | Mfr: KP9004A| Item: 0457878

Polish the surfaces of your crucibles and molds to maintain high standards in your sample preparation process

$2,504.76CAD / Each

$2,504.76CAD / Each
Product Description

Includes

Electric handheld rotary tool, flexible shaft, table stand, sets of buffing pads, and diamond pastes of assorted grades.

Key Features

  • Restore the shiny finish to platinumware of any shape and size
  • For 115 VAC operation

More About this Item

Proper maintenance of your platinumware will extend the service life as well as ensure great results.

Specifications
  • Compatible WithAll types of platinumware
  • Power (VAC)115
  • Power (Hz)60
  • DescriptionPlatinumware Polishing Kit; 115 VAC, 60 Hz
Reviews
Q&A