Environmental Express Electrode pH submersible 25 ft cable length BNC
Representative image only.
Environmental Express Electrode pH submersible 25 ft cable length BNC
Environmental Express | Item: 3580552
Ideal for field applications
$331.07CAD / Each
BUY MORE & SAVE
  • 1-3$331.07 CAD / Each
  • 4+$297.96 CAD / Each
$331.07CAD / Each
BUY MORE & SAVE
  • 1-3$331.07 CAD / Each
  • 4+$297.96 CAD / Each
Product Description
Key Features
  • Completely submersible up to the last one foot prior to the meter connection
  • Extended cable lengths up to 100 feet
  • 1 year warranty
More About this Item
These ABS plastic electrodes are designed for field applications. Annular junction provides fast response and resists pressure effects of submersion. BNC connection is compatible with most pH meters. 100 foot cable length electrode includes preamplification.
1 Year Warranty
Specifications
  • Electrode Body MaterialEpoxy
  • Min pH (pH)0
  • Max pH (pH)12
  • Min Temperature (° C)0
  • Max Temperature (° C)60
  • ConnectorBNC
  • DescriptionElectrode pH submersible 25 ft cable length BNC