Environmental Express EC 2000 Benchtop Meter Kit

Representative image only.

Environmental Express EC 2000 Benchtop Meter Kit

Environmental Express | Item: 3541960

Developed using extensive feedback from laboratory staff, our new Environmental Express 2000 series meters are assembled in various configurations for measuring pH/mV, EC, Ion, DO, BOD parameters

$1,968.45CAD / Each

$1,968.45CAD / Each

Buy It Together

Current Item

Environmental Express EC 2000 Benchtop Meter Kit
Environmental Express EC 2000 Benchtop Meter Kit
3541960

$1,968.45CAD / Each
+
Environmental Express 2000 All-in-One pH Electrode
3541952

$282.89CAD / Each
+
Environmental Express pH 2000 Benchtop Meter Kit
3541954

$1,701.77CAD / Each
+ + =
Total Price 6446.97 ADD 5 ITEMS TO CART
Product Description

Key Features

  • Various configurations available to meet the demands of measuring pH/mV, EC, Ion, PC, PD, DO, BOD parameters
  • Meter Kit formats include probes, 35419-54, -60, -65, -74, -78
  • Automatic record retention and accessibility for continuous compliance with regulatory requirements
  • 0.01, 0.001 resolution for routine or research applications where acute accuracy is critical
  • Large displays with a small footprint, ideal for labs with limited bench space
  • Automatic functions like buffer recognition and auto calibration for ease of use at any skill level

More About this Item

Environmental Express provides a diverse range of water quality analyzers and electrodes that are designed specifically for laboratory applications with compliance in mind. Our multi-parameter benchtops include functionality to meet the highly specific and/or broad ranging needs of wastewater, drinking water, food and beverage, chemical and pharmaceutical testing labs.

These benchtop meters offer a high-capacity internal memory that can capture and store 2000 data points with the time and date. They have a time-based auto data log with GLP compliant real time clocks. This data can be exported into CSV, Excel, or PDF formats via USB cable and use of complimentary software. A password access protection function secures the meter setup mode from unauthorized access. An adjustable shutoff time up to 30 minutes turns the meter off when idle.

Specifications
  • Meter TypeConductivity/Resistivity/TDS/Salinity/Temp (°C/°F)
  • Display type5” Custom LCD with backlight and 320 segments
  • ConnectionsBNC, phono, DC sockets
  • Min Conductivity0.000 µS/cm (k = 0.1)
  • Max Conductivity2 S/cm (k=10)
  • Conductivity Resolution0.05% full scale
  • Conductivity Accuracy± 0.6% full scale; ± 1.5% full scale > 18.0 mS/cm
  • Min TDS0.01 mg/L (ppm)
  • Max TDS100 g/L (ppt)
  • TDS Resolution0.01, 0.1, 1 mg/L - g/L (ppm - ppt)
  • TDS Accuracy± 0.1% full scale
  • Min Salinity0.00 ppt / 0.00%
  • Max Salinity100.0 ppt / 10.00%
  • Salinity Resolution0.1 ppt / 0.01%
  • Salinity Accuracy± 0.2% full scale
  • Min Resistivity0.000 Ω•cm
  • Max Resistivity20.0 MΩ•cm
  • Resistivity Resolution0.5% full scale
  • Resistivity Accuracy± 0.6% full scale; ± 1.5% full scale > 1.80 MΩ•cm
  • Min Temperature (° C)-30
  • Max Temperature (° C)130
  • Min Temperature (° F)-22
  • Max Temperature (° F)266
  • Temperature Resolution0.1 °C / °F
  • Temperature Accuracy± 0.5 °C / ± 0.9 °F
  • OutputPhono jack (USB/RS-232C)
  • Data Logging (points)2000
  • Power (VAC)100 to 240
  • Power (Hz)50-60 Hz
  • Length (cm)15.5
  • Width (cm)15
  • Height (cm)6.7
  • Width (in)5 29/32
  • Length (in)6 7/64
  • Height (in)2 41/64
  • DescriptionEC 2000 Benchtop Meter Kit
  • Warranty3 years
Reviews
Q&A