Environmental Express Buffer Solution, pH 2.00; 1000 mL

Representative image only.

Environmental Express Buffer Solution, pH 2.00; 1000 mL

Environmental Express | Item: 0065576

Calibrate pH instruments to ensure accuracy

$46.41CAD / Each

$46.41CAD / Each
Product Description

Key Features

  • Lab-grade buffers available in multiple volumes to reduce waste and support various levels of throughput
  • Color coded and/or clearly labeled for easy identification in busy labs
  • Unopened, shelf life is 2 years
  • NIST/USA buffer value of pH 2.00

More About this Item

pH 2.00 is a low-range pH meter calibration point. All Environmental Express® pH buffer solutions are manufactured under ISO 9000 quality guidelines and traceable to NIST standard (for meters with 0.01 resolution). A pH/temperature table is printed on the label for accurate calibrations.

If unopened, shelf life of pH 2.00 is 2 years; once opened, use within 90 days.

Store buffers away from direct sunlight.

Specifications
  • Buffer Value2
  • Volume (mL)1000
  • ColorClear
  • DescriptionBuffer Solution, pH 2.00; 1000 mL
Reviews
Q&A