Endecotts ZSICSA-.500 Sieve Calibration Standard, soda-lime glass, 35 mesh
Representative image only.
Endecotts ZSICSA-.500 Sieve Calibration Standard, soda-lime glass, 35 mesh
Endecotts | Mfr: ZSICSA-.500| Item: 5994509
Spheres are traceable to the United Kingdom National Physical Laboratory
$340.35CAD / Each
$340.35CAD / Each
Product Description
Key Features
  • Calibrate sieves to an accuracy of ±1µm
  • 6 month warranty
More About this Item
Soda-lime glass microspheres pass over the entire surface of the sieve and through every aperture. Standards include a calibration chart.
6 Month WarrantyISO9001 Certified Supplier
Specifications
  • U.S. Standard Screen Size (No.)35
  • Tyler Standard Screen Size (mesh)32
  • Min Sieve Aperture (microns)425
  • Max Sieve Aperture (microns)600
  • Weight (g)12.5
  • DescriptionSieve Calibration Standard, soda-lime glass, 35 mesh
  • Warranty6 Months