Endecotts ZSICSA-.212 Sieve Calibration Standard, soda-lime glass, 70 mesh
Representative image only.
Endecotts ZSICSA-.212 Sieve Calibration Standard, soda-lime glass, 70 mesh
Endecotts | Mfr: ZSICSA-.212| Item: 5994514
Spheres are traceable to the United Kingdom National Physical Laboratory
$534.10CAD / Each
$534.10CAD / Each
Product Description
Key Features
  • Calibrate sieves to an accuracy of ±1µm
  • 6 month warranty
More About this Item
Soda-lime glass microspheres pass over the entire surface of the sieve and through every aperture. Standards include a calibration chart.
6 Month WarrantyISO9001 Certified Supplier
Specifications
  • U.S. Standard Screen Size (No.)70
  • Tyler Standard Screen Size (mesh)65
  • Min Sieve Aperture (microns)180
  • Max Sieve Aperture (microns)250
  • Weight (g)7.5
  • DescriptionSieve Calibration Standard, soda-lime glass, 70 mesh
  • Warranty6 Months