Endecotts ZMWSK-OCT200/8 Wet Sieving Kit for Octagon 200 & Octagon 200CL with 8" Sieves
Representative image only.
Endecotts ZMWSK-OCT200/8 Wet Sieving Kit for Octagon 200 & Octagon 200CL with 8" Sieves
Endecotts | Mfr: ZMWSK-OCT200/8| Item: 5888683
Optimize your sieving with a wide range of equipment
$1,944.87CAD / Each
$1,944.87CAD / Each
Product Description
Includes
Top clamping plate with acrylic cover and spray rose, watertight O-ring seals, and a stainless steel reciever with drainage spout
Key Features
  • Wet sieving conversion kits allow you to sieve wet material
  • SieveWare software automatically controls the measurement and weighing procedures
  • 2 year warranty
More About this Item
These accessories ensure that Endecotts sieve shakers can function in a variety of applications.
Two year warranty
Specifications
  • Compatible WithOctagon 200 & 200CL
  • DescriptionWet Sieving Kit for 8" Sieves
  • Warranty2 Years