Eldon James Hose Barb Fitting, Y Union, Natural Kynar®, 3/16" ID; 10/PK
Representative image only.
Eldon James Hose Barb Fitting, Y Union, Natural Kynar®, 3/16" ID; 10/PK
Eldon James | Mfr: Y0-3NK7| Item: 5012899
Excellent for high temperature and harsh physical environments
$55.40CAD / Pkg of 10
$55.40CAD / Pkg of 10
Product Description
Key Features
  • Ideal for general-purpose, laboratory, and single-use applications to connect tubing with the rest of your system
  • Free technical application support available to determine which connector type is needed for your application
  • PVC-free—ideal for use in bioprocess, life science, and pharmaceutical industries
  • Quick and easy assembly without tools
  • Biopharma compatible and USP 66
  • BPOG compliant
  • 3/16" hose barb
More About this Item
Use these high-quality PVC-free fittings in biomedical research, pharmaceutical, electronic, and other general-purpose applications. Easily combine or split fluid flow using Y unions. Each fitting features a single barb on each end, ensuring a tight seal to keep the tubing securely in place.
An anti-rotation device on each port prevents tubing from rotating, minimizing kinks. These natural Kynar® fittings provide excellent chemical, flame, gamma radiation, UV, and weather resistance. They also exhibit good mechanical properties, low moisture absorption, and good machinability.
Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.
Specifications
  • Fitting MaterialPVDF;PVDF
  • Sterilization MethodE-beam/gamma irradiation, EtO, or autoclave up to 135°C;E-beam/gamma irradiation, EtO, or autoclave up to 135°C
  • Number Of Ports3;3
  • Connection TypeHose Barb Y;Hose Barb Y
  • Barb Size / Tubing Id (Port 1) (in)3/16;3/16
  • Barb Size / Tubing Id (Port 2) (in)3/16;3/16
  • Barb Size / Tubing Id (Port 3) (in)3/16;3/16
  • Flow PatternY;Y
  • ColorNatural;Natural
  • Min Temperature (° F)-31;-31
  • Max Temperature (° F)275;275
  • Min Temperature (° C)-35;-35
  • Max Temperature (° C)135;135
  • Regulatory ApprovalsUSP;USP;Non-Phthalate;Non-DEHP;Non-DEHP;Non-Phthalate
  • DescriptionHose Barb Fitting, Y Union, Natural Kynar®, 3/16" ID; 10/PK;Hose Barb Fitting, Y Union, Natural Kynar®, 3/16" ID; 10/PK