Edwards Ultragrade 15 Performance Oil, 1 L Bottle
Representative image only.
Edwards Ultragrade 15 Performance Oil, 1 L Bottle
Edwards | Mfr: H11026015| Item: 7930053
Enhance the performance of your vacuum pumps with the right vacuum pump oil
$91.20CAD / Each
$91.20CAD / Each
Product Description
Key Features
  • Specifically designed for vacuum pumps and applications
  • Combined with additives to offer improved vacuum performance, thermal and oxidation stability, and rust inhibition
  • Provides excellent lubrication, reducing wear and extending the lifespan of pump components
  • 1 L bottle
  • 1 year warranty
More About this Item
Select the right pump oil for your vacuum pumps to keep your equipment in excellent working condition.
Maximize the performance and longevity of your vacuum pump with Edwards Ultragrade 15 High-Performance Vacuum Pump Oil. This premium-grade lubricating oil is specially formulated to enhance vacuum efficiency, minimize wear, and provide exceptional thermal stability in rotary vane vacuum pumps. Designed for small rotary vane pumps (0.5 to 5 m3/hr), it ensures low vapor pressure, superior lubrication, and extended service life, reducing the need for frequent maintenance.
1 Year Warranty
Specifications
  • MaterialUltragrade 15 mineral oil
  • Compatible WithSmall rotary vane pumps (0.5-5m3/hr) where cold start capability or low starting torque is needed
  • Quantity1 L
  • DescriptionUltragrade 15 Performance Oil, 1 L