Dynalon PTFE Joint Sleeves for Standard Taper Size 34/45; 6/Pk
Representative image only.
Dynalon PTFE Joint Sleeves for Standard Taper Size 34/45; 6/Pk
Dynalon | Mfr: 362655| Item: 0607345
$59.52CAD / Pkg of 6
BUY MORE & SAVE
  • 1-5$59.52 CAD / Pkg of 6
  • 6-29$53.57 CAD / Pkg of 6
  • 30-119$50.59 CAD / Pkg of 6
  • 120+$47.62 CAD / Pkg of 6
$59.52CAD / Pkg of 6
BUY MORE & SAVE
  • 1-5$59.52 CAD / Pkg of 6
  • 6-29$53.57 CAD / Pkg of 6
  • 30-119$50.59 CAD / Pkg of 6
  • 120+$47.62 CAD / Pkg of 6
Product Description
Key Features
  • 1 year warranty
More About this Item
Translucent PTFE, self-lubricating sleeves are precision tapered for a snug fit. Withstand temperatures from –103 to 482°>F (–75 to 250°>C). Pack of 6. 3 pk/cs*.
1 Year Warranty
Specifications
  • Taper Size34/45
  • MaterialPolytetrafluoroethylene (PTFE)
  • DescriptionPTFE Joint Sleeves for Standard Taper Size 34/45; 6/Pk