Dynalon PTFE Joint Sleeves for Standard Taper Size 19/38; 6/Pk
Representative image only.
Dynalon PTFE Joint Sleeves for Standard Taper Size 19/38; 6/Pk
Dynalon | Mfr: 362625| Item: 0607338
$42.43CAD / Pkg of 6
BUY MORE & SAVE
  • 1-5$42.43 CAD / Pkg of 6
  • 6-29$38.19 CAD / Pkg of 6
  • 30-119$36.07 CAD / Pkg of 6
  • 120+$33.94 CAD / Pkg of 6
$42.43CAD / Pkg of 6
BUY MORE & SAVE
  • 1-5$42.43 CAD / Pkg of 6
  • 6-29$38.19 CAD / Pkg of 6
  • 30-119$36.07 CAD / Pkg of 6
  • 120+$33.94 CAD / Pkg of 6
Product Description
Key Features
  • 1 year warranty
More About this Item
Translucent PTFE, self-lubricating sleeves are precision tapered for a snug fit. Withstand temperatures from –103 to 482°>F (–75 to 250°>C). Pack of 6. 3 pk/cs*.
1 Year Warranty
Specifications
  • Taper Size19/38
  • MaterialPolytetrafluoroethylene (PTFE)
  • DescriptionPTFE Joint Sleeves for Standard Taper Size 19/38; 6/Pk