Dynalon Polypropylene Buchner funnel, 110 mm dia
Representative image only.
Dynalon Polypropylene Buchner funnel, 110 mm dia
Dynalon | Mfr: 242835-110| Item: 0612805
Get quick, economical vacuum filtration
$76.17CAD / Each
BUY MORE & SAVE
  • 1-5$76.17 CAD / Each
  • 6-29$68.55 CAD / Each
  • 30-119$64.74 CAD / Each
  • 120+$60.94 CAD / Each
$76.17CAD / Each
BUY MORE & SAVE
  • 1-5$76.17 CAD / Each
  • 6-29$68.55 CAD / Each
  • 30-119$64.74 CAD / Each
  • 120+$60.94 CAD / Each
Buy It Together
Current Item
Dynalon Polypropylene Buchner funnel, 110 mm dia
Dynalon Polypropylene Buchner funnel, 110 mm dia
0612805
$76.17CAD / Each
+ =
Total Price 109.76 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Easy sterilization: HDPE by gas, PP by autoclave or gas
  • 1 year warranty
More About this Item
Choose Buchner funnels for gravity or vacuum filtration. Recommended sterilization: HDPE for gas, PP by autoclave or gas.
1 Year Warranty
Specifications
  • MaterialPolypropylene (PP)
  • Filter Paper Diameter (mm)110
  • DescriptionPolypropylene Buchner funnel, 110 mm dia