Dynalon Chemical Bottle Pourer, PTFE, 38-mm cap size
Representative image only.
Dynalon Chemical Bottle Pourer, PTFE, 38-mm cap size
Dynalon | Mfr: 310374-0004| Item: 0603804
$165.02CAD / Pkg of 1
BUY MORE & SAVE
  • 1-4$165.02 CAD / Pkg of 1
  • 5-9$148.52 CAD / Pkg of 1
  • 10+$132.02 CAD / Pkg of 1
$165.02CAD / Pkg of 1
BUY MORE & SAVE
  • 1-4$165.02 CAD / Pkg of 1
  • 5-9$148.52 CAD / Pkg of 1
  • 10+$132.02 CAD / Pkg of 1
Product Description
Key Features
  • 1 year warranty
More About this Item
These worry-free polytetrafluoroethylene (PTFE) pourers have inert pouring surfaces — safely pour hazardous and corrosive chemicals. A breather vent allows direct and controlled pouring. Tethered strap keeps closure handy. Notch in tether keeps closure out of the way when pouring. A PTFE-encapsulated sealing ring ensures that bottle contents remain contaminant free. Fit standard reagent bottles; order bottles separately.
1 Year Warranty
Specifications
  • MaterialPolytetrafluoroethylene (PTFE)
  • Cap MaterialPolytetrafluoroethylene (PTFE)
  • Cap / Stopper SizeGL38
  • Cap ColorBlue
  • DescriptionChemical Bottle Pourer, PTFE, 38-mm cap size