Dynalon 797303-0030 Bio-bin Waste Disposal Container, 30L, Case of 10

Representative image only.

Dynalon 797303-0030 Bio-bin Waste Disposal Container, 30L, Case of 10

Dynalon | Mfr: 797303-0030| Item: 8140209

Great alternative to plastic waste collection and disposal containers

$360.61CAD / Case of 10

$360.61CAD / Case of 10

Buy It Together

Current Item

Dynalon 797303-0030 Bio-bin Waste Disposal Container, 30L, Case of 10
Dynalon 797303-0030 Bio-bin Waste Disposal Container, 30L, Case of 10
8140209

$360.61CAD / Case of 10
+ =
Total Price 629.14 ADD 2 ITEMS TO CART
Product Description

Includes

2" x 2" "biohazard" waste labels.

Key Features

  • Space-efficient, lightweight fold-flat design
  • Waterproof construction — inside and outside — with heat-sealed joints
  • Built-in super absorbent Bio-matt™ liner soaks up excess liquid

More About this Item

These disposable plastic-coated paper containers are perfect for laboratory waste that requires special heat treatment such as autoclaving or incineration before disposal. The built-in lid has both a temporary and permanent seal and features a handle for easy transport when full. Ideal for use in academic, clinical, government, and industrial laboratories for a wide range of non-sharp biohazard or non-hazardous waste. Autoclavable

This new floor size bio-bin is ideal for use in labs, wards and for community health. Space efficient fold-flat design with easy assembly. Reduce waste disposal costs and carbon footprint. Built-in Bio-matt absorbs approxmately 1/2 liter of liquid.

Specifications
  • Capacity (Liters)30
  • Length (in)11
  • Width (in)11
  • Length (cm)27.94
  • Width (cm)27.94
  • DescriptionBio-bin Waste Disposal Container, 30L, Case of 10
Reviews
Q&A