Dynalon 797303-0006 Bio-bin Waste Disposal Container, 6L, Case of 40

Representative image only.

Dynalon 797303-0006 Bio-bin Waste Disposal Container, 6L, Case of 40

Dynalon | Mfr: 797303-0006| Item: 8140208

Great alternative to plastic waste collection and disposal containers

$486.22CAD / Case of 40

$486.22CAD / Case of 40

Buy It Together

Current Item

Dynalon 797303-0006 Bio-bin Waste Disposal Container, 6L, Case of 40
Dynalon 797303-0006 Bio-bin Waste Disposal Container, 6L, Case of 40
8140208

$486.22CAD / Case of 40
+ =
Total Price 563.57 ADD 2 ITEMS TO CART
Product Description

Includes

2" x 2" "biohazard" waste labels.

Key Features

  • Space-efficient, lightweight fold-flat design
  • Waterproof construction — inside and outside — with heat-sealed joints
  • Built-in super absorbent Bio-matt™ liner soaks up excess liquid

More About this Item

These disposable plastic-coated paper containers are perfect for laboratory waste that requires special heat treatment such as autoclaving or incineration before disposal. The built-in lid has both a temporary and permanent seal and features a handle for easy transport when full. Ideal for use in academic, clinical, government, and industrial laboratories for a wide range of non-sharp biohazard or non-hazardous waste. Autoclavable

The 6 liter Bio-bin is specially designed for the collection and disposal of contaminated laboratory serological pipettes and other long, non-sharps items up to 15" (380mm) in length. The 6 liter Bio-bins are available with two (2) styles of stainless steel stands for bench/work station (U Stand) and floor use (Z Stand). The U Stand conveniently holds the Bio-bin in place at an angle and is ideal for use in biological safety cabinets.

Specifications
  • Capacity (Liters)6
  • Length (in)6
  • Width (in)6
  • Length (cm)15.24
  • Width (cm)15.24
  • DescriptionBio-bin Waste Disposal Container, 6L, Case of 40
Reviews
Q&A