Dwyer 475-6-FM Digital Manometer, 30 psi

Representative image only.

Dwyer 475-6-FM Digital Manometer, 30 psi

Dwyer | Mfr: 475-6-FM| Item: 6806276

Measures positive, negative and differential pressures

$491.38CAD / Each

Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

$491.38CAD / Each

Buy It Together

Current Item

Dwyer 475-6-FM Digital Manometer, 30 psi
Dwyer 475-6-FM Digital Manometer, 30 psi
6806276

$491.38CAD / Each
+ + =
Total Price 943.71 ADD 3 ITEMS TO CART
Product Description

Includes

one 9V battery.

Key Features

  • ±0.5% full scale accuracy
  • Rugged, extruded aluminum case
  • Selectable english/metric units
  • 1 year warranty

More About this Item

The Dwyer® Series 475 Mark III handheld digital manometer is ideal for field calibration, monitoring or trouble shooting HVAC systems, clean rooms or a wide range of other low pressure pneumatic systems. This handy instrument measures positive, negative or differential pressures of air and natural gases in ranges from 1" WC to 150 psi. The manometer is approved and is intrinsically safe for hazardous locations, Class 1, Div.1, Group A, B,C, D, T4. Its simple operation and easy to read digital display make it an indispensable test instrument for the plant engineer, industrial hygienist and HVAC technician. The manometer is housed in a durable extruded aluminum case with its solid state circuitry mounted on a tough fiberglass epoxy circuit board. To meet the most demanding applicat

1 Year Warranty
Specifications
  • Accuracy±0.5% full-scale (60 to 78°F); ±1.5% full-scale below 60°F and above 78°F
  • Min Pressure (PSI)0
  • Max Pressure (PSI)30
  • Min Temperature (° F)32
  • Max Temperature (° F)104
  • Min Temperature (° C)0
  • Max Temperature (DegreeC)40
  • CaseAluminum
  • Process Connection1/8" NPT(F)
  • Unit Of Measurementpsi and bar
  • Display4-digit LCD, 0.42"H
  • Battery Type1 9V (included)
  • Battery Life100 hours
  • Media CompatibilityNoncorrosive gases
  • Resolution0.01 psi
  • Depth (in)1 3/4
  • Height (in)7
  • Width (in)2 1/2
  • Depth (cm)4.4
  • Height (cm)17.8
  • Width (cm)6.4
  • DescriptionDigital Manometer, 30 psi
  • Warranty1 year
Reviews
Q&A