DWK Life Sciences (Wheaton) Tubular Serum Vials, European Blowback, ISO 8362-1, Clear Glass, 6R, 10 mL; 100/CS

Representative image only.

DWK Life Sciences (Wheaton) Tubular Serum Vials, European Blowback, ISO 8362-1, Clear Glass, 6R, 10 mL; 100/CS

DWK Life Sciences (Wheaton) | Mfr: WEBB06R-T-EP| Item: 0891629

Shrink-wrapped modules reduce particulate contamination

$161.34CAD / Case of 100

$161.34CAD / Case of 100
Product Description

Key Features

  • Made from Type I borosilicate glass tubing
  • Vials conform to requirements outlined in ISO 8362 EP
  • Serum vial feature 10 mL brimful capacity in size 6R
  • Vials are packaged 10 rows of 10 tubes per tray

More About this Item

These WHEATON® tubular serum vials with European blowback, conform to ISO 8362 EP standards, and are constructed of Type I borosilicate glass. The blowback feature in the neck and specially designed bottom radius for added strength are important for lyophilization applications. Vials are packaged in shrink wrapped trays to greatly reduce particulate contamination.

Specifications
  • Volume (mL)10
  • Tube MaterialGlass
  • Tube Bottom ShapeFlat
  • Tube ColorClear
  • Cap TypeNot Included
  • SterileNo
  • Height (in)1 19/32
  • Outside Diameter (in)29/32
  • Height (mm)40
  • Outside Diameter (mm)22
  • Special FeaturesSize 6R
  • DescriptionTubular Serum Vials, European Blowback, ISO 8362-1, Clear Glass, 6R, 10 mL; 100/CS
Reviews
Q&A