DWK Life Sciences (Wheaton) Tubular Serum Vials, European Blowback, ISO 8362-1, Clear Glass, 10R, 13.5 mL; 150/CS

Representative image only.

DWK Life Sciences (Wheaton) Tubular Serum Vials, European Blowback, ISO 8362-1, Clear Glass, 10R, 13.5 mL; 150/CS

DWK Life Sciences (Wheaton) | Mfr: WEBB10R-T-EP| Item: 0891630

Shrink-wrapped modules reduce particulate contamination

$274.05CAD / Case of 150

$274.05CAD / Case of 150
Product Description

Key Features

  • Made from Type I borosilicate glass tubing
  • Vials conform to requirements outlined in ISO 8362 EP
  • Serum vial feature 13.5 mL brimful capacity in size 10R
  • Vials are packaged 6 rows of 13 tubes and 6 rows of 12 tubes per tray

More About this Item

These WHEATON® tubular serum vials with European blowback, conform to ISO 8362 EP standards, and are constructed of Type I borosilicate glass. The blowback feature in the neck and specially designed bottom radius for added strength are important for lyophilization applications. Vials are packaged in shrink wrapped trays to greatly reduce particulate contamination.

Specifications
  • Volume (mL)13.5
  • Tube MaterialGlass
  • Tube Bottom ShapeFlat
  • Tube ColorClear
  • Cap TypeNot Included
  • SterileNo
  • Height (in)1 51/64
  • Outside Diameter (in)29/32
  • Height (mm)45
  • Outside Diameter (mm)24
  • Special FeaturesSize 10R
  • DescriptionTubular Serum Vials, European Blowback, ISO 8362-1, Clear Glass, 10R, 13.5 mL; 150/CS
Reviews
Q&A