DWK Life Sciences (Wheaton) Serum Vials, US Blowback, EP 3.2.1, Amber Glass, 5 mL; 100/CS

Representative image only.

DWK Life Sciences (Wheaton) Serum Vials, US Blowback, EP 3.2.1, Amber Glass, 5 mL; 100/CS

DWK Life Sciences (Wheaton) | Mfr: WUBB5MLA-T-EP| Item: 0891625

Special design adds strength for freeze-drying applications

$177.91CAD / Case of 100

$177.91CAD / Case of 100
Product Description

Key Features

  • Made from Type I borosilicate glass tubing
  • Vials conform to the hydrolytic resistance requirements outlined in USP <660> and EP 3.2.1
  • Shrink-wrapped modules reduce particulate contamination
  • Serum vial features 5 mL capacity
  • Vial is constructed of amber glass for light sensitive samples

More About this Item

These WHEATON® serum vials with US blowback are constructed of Type I borosilicate glass, and available in clear or amber colors. The blowback feature in the neck and specially designed bottom radius for added strength are important for lyophilization applications. Vials are packaged in shrink wrapped trays to greatly reduce particulate contamination.

Specifications
  • Volume (mL)5
  • Tube MaterialGlass
  • Tube Bottom ShapeFlat
  • Tube ColorAmber
  • Cap TypeNot Included
  • SterileNo
  • Height (in)1 19/32
  • Outside Diameter (in)55/64
  • Height (mm)40
  • Outside Diameter (mm)21.75
  • DescriptionSerum Vials, US Blowback, EP 3.2.1, Amber Glass, 5 mL; 100/CS
Reviews
Q&A