DWK Life Sciences (Wheaton) Serum Vials, Clear Type I Glass, 20 mL; 120/Cs

Representative image only.

DWK Life Sciences (Wheaton) Serum Vials, Clear Type I Glass, 20 mL; 120/Cs

DWK Life Sciences (Wheaton) | Mfr: 223687| Item: 0891604

Special design adds strength for freeze drying applications

$269.34CAD / Case of 120

$269.34CAD / Case of 120
Product Description

Key Features

  • Made from low extractable Type 1 borosilicate glass

More About this Item

The blowback feature in the neck and specially designed bottom radius for added strength
are important for lyophilization applications. Vials are packaged in shrink wrapped modules
to greatly reduce particulate contamination. Available in clear or amber glass. Clear vials
also conform to ASTM Type I and Class A requirements.

Specifications
  • Volume (mL)20
  • Tube MaterialBorosilicate glass
  • Tube Bottom ShapeFlat
  • Tube ColorClear
  • Height (in)2 9/32
  • Outside Diameter (in)1 1/4
  • Height (mm)58
  • Outside Diameter (mm)30.5
  • DescriptionSerum Vials, Clear Type I Glass, 20 mL; 120/Cs
Reviews
Q&A