DWK Life Sciences (Wheaton) Coplin Staining Jar with Cover, Glass, 8-16 Slides

Representative image only.

DWK Life Sciences (Wheaton) Coplin Staining Jar with Cover, Glass, 8-16 Slides

DWK Life Sciences (Wheaton) | Mfr: 900620| Item: 4858520

Ideal for staining slides

$114.19CAD / Each

$114.19CAD / Each

Buy It Together

Current Item

DWK Life Sciences (Wheaton) Coplin Staining Jar with Cover, Glass, 8-16 Slides
DWK Life Sciences (Wheaton) Coplin Staining Jar with Cover, Glass, 8-16 Slides
4858520

$114.19CAD / Each
+ =
Total Price 388.24 ADD 2 ITEMS TO CART

Customers Also Bought

Product Description

Key Features

  • Constructed of durable soda-lime glass
  • Jar is designed to hold 3” x 1” (75mm x 25mm) slides
  • Jar holds 8 single 3” x 1” (75mm x 25mm) slides vertically or 16 slides back-to-back

More About this Item

WHEATON® Coplin staining jars provide a stable container for staining slides. Glass covers protect slides from outside elements and reduce reagent evaporation. Compact size makes it easy to transfer slides and requires only a small volume of reagent.

Specifications
  • Capacity (Number of slides)8 single or 16 back-to-back
  • Internal Width (in)1
  • Internal Height (in)3 19/64
  • Internal Depth (in)1 27/32
  • Internal Width (cm)2.6
  • Internal Height (cm)8.5
  • Internal Depth (cm)4.7
  • DescriptionCoplin Staining Jar with Cover, Glass, 8-16 Slides
Reviews
Q&A