DWK Life Sciences (Wheaton) CompletePAK® Sterile Vial Kit, 2 mL, Ultra Pure Stopper, Blue; 210/Cs

Representative image only.

DWK Life Sciences (Wheaton) CompletePAK® Sterile Vial Kit, 2 mL, Ultra Pure Stopper, Blue; 210/Cs

DWK Life Sciences (Wheaton) | Mfr: W991000-CP| Item: 0891819

Reduce scrap costs—equal number of vials, stoppers, and seals

$3,637.80CAD / Case of 210

$3,637.80CAD / Case of 210

Buy It Together

Current Item

DWK Life Sciences (Wheaton) CompletePAK® Sterile Vial Kit, 2 mL, Ultra Pure Stopper, Blue; 210/Cs
DWK Life Sciences (Wheaton) CompletePAK® Sterile Vial Kit, 2 mL, Ultra Pure Stopper, Blue; 210/Cs
0891819

$3,637.80CAD / Case of 210
+ =
Total Price 5099.40 ADD 2 ITEMS TO CART
Product Description

Includes

2 mL clear-glass sterile vials, 13 mm sterile Ultra Pure serum stoppers, sterile blue seals; case of 210

Key Features

  • Vial packs have sustained particulate cleaning, depryogenation, and sterilization
  • Ready to use—right out of the box
  • 18-month shelf life from date of processing
  • 2 mL vials, 13 mm ultra-pure stoppers, and blue seals
  • Ultra-pure stoppers are made from an ultra pure bromobutyl formulation with extremely high chemical purity and low gas permeability
  • Compatible with applications such as Water for Injection (WFI) where rubber extractables can cause problems

More About this Item

CompletePAK® vial kits include everything you need, without scrap costs, and no difference in component lead times. These clear-glass vials come with stoppers and seals—choose the volume, cap size and type, and seal color to suit your needs.

These presterilized vial kits are ready to use right off the shelf. Processing includes particulate cleaning, depryogenation, and sterilization—ideal for drug research, freeze drying/lyophilization, aseptic filling, and large/small molecule packaging.

NonpyrogenicSterile
Specifications
  • Volume (mL)2
  • Tube MaterialGlass
  • Tube Bottom ShapeFlat
  • Tube ColorClear
  • Cap TypeUltra pure, standard
  • Cap Diameter (mm)13
  • Cap colorBlue
  • SterileYes
  • AutoclavableYes
  • DescriptionCompletePAK® Sterile Vial Kit, 2 mL, Ultra Pure Stopper, Blue; 210/Cs
Reviews
Q&A