DWK Life Sciences (Wheaton) CompletePAK® Sterile Serum Vial, 5 mL; 200/Cs

Representative image only.

DWK Life Sciences (Wheaton) CompletePAK® Sterile Serum Vial, 5 mL; 200/Cs

DWK Life Sciences (Wheaton) | Mfr: W987001-S| Item: 0891879

Order individual system components—pre-sterilized!

$717.54CAD / Case of 200

$717.54CAD / Case of 200
Product Description

Key Features

  • Keep a spare or order components separately to buil your own system
  • Sterile, clear-glass vial, 5 mL capacity

More About this Item

Separate vial components (vials, stoppers, and seals) come in pre-sterilized cases. Order a spare, or build-your-own vial system.

Sterile
Specifications
  • Volume (mL)5
  • Tube MaterialGlass
  • Tube Bottom ShapeFlat
  • Tube ColorClear
  • Cap TypeNot applicable
  • Cap Diameter (mm)20
  • Cap colorNot applicable
  • SterileYes
  • AutoclavableYes
  • DescriptionCompletePAK® Sterile Serum Vial, 5 mL; 200/Cs
Reviews
Q&A