DWK Life Sciences (Kimble) KC20028W-500 Class A, Graduated Cylinders with Bumpers, 500 mL

Representative image only.

DWK Life Sciences (Kimble) KC20028W-500 Class A, Graduated Cylinders with Bumpers, 500 mL

DWK Life Sciences (Kimble) | Mfr: KC20028W-500| Item: 3452748

$307.94CAD / Case of 1

for an additional
$155.44CAD / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services

$307.94CAD / Case of 1

Buy It Together

Current Item

DWK Life Sciences (Kimble) KC20028W-500 Class A, Graduated Cylinders with Bumpers, 500 mL
DWK Life Sciences (Kimble) KC20028W-500 Class A, Graduated Cylinders with Bumpers, 500 mL
3452748

$307.94CAD / Case of 1
+ =
Total Price 463.38 ADD 2 ITEMS TO CART
Product Description

Key Features

  • Calibrated “to deliver”

More About this Item

Ultra-clear safety-coated glassware helps stop and control spills in the event of glassware breakage. If a break does occur, the durable odor-free glassware will reduce the hazards of shattered glass and leakage of toxic or corrosive chemicals. Glassware has a unique nonslip surface when wet or dry.Class A graduated cylinders are unique with reverse metric scale with white markings. Cylinders include a bumper and a ground base for stability. Autoclavable to120°C (250°F).

Specifications
  • MaterialGlass
  • Capacity (mL)500
  • Graduation Interval (mL)5
  • Tolerance (mL)±1.3
  • DescriptionClass A, Graduated Cylinders with Bumpers, 500 mL
Reviews
Q&A