DWK Life Sciences (Kimble) HPLC Mobile Phase Reservoir, Glass, Conical Bottom; 0.5 Liter
Representative image only.
DWK Life Sciences (Kimble) HPLC Mobile Phase Reservoir, Glass, Conical Bottom; 0.5 Liter
DWK Life Sciences (Kimble) | Mfr: 953901-0502| Item: 0201700
Chemically inert glass prevents leaching of any extractables into the mobile phase solvents
$421.76CAD / Each
$421.76CAD / Each
Product Description
Key Features
  • Constructed of durable safety coated glass
  • 1 year warranty
More About this Item
These reservoirs are manufactured of high-quality borosilicate glass for maximum chemical resistance. Reservoirs are coated with plastic for safety and UV resistance. Select from conical bottom configuration for maximum solvent utilization or flat bottom reservoirs.
1 Year Warranty
Specifications
  • Bottom TypeConical
  • Total Volume Capacity (mL)500
  • DescriptionHPLC Mobile Phase Reservoir, Glass, Conical Bottom; 0.5 Liter