DWK Life Sciences (Kimble) 73770-20125 Disposable Round Bottom Culture Tubes, 20 x 125, no markings, 500/cs
Representative image only.
DWK Life Sciences (Kimble) 73770-20125 Disposable Round Bottom Culture Tubes, 20 x 125, no markings, 500/cs
DWK Life Sciences (Kimble) | Mfr: 73770-20125| Item: 3458058
Ideal for general bacteriological and tissue culture applications
$670.02CAD / Case of 500
BUY MORE & SAVE
  • 1-4$670.02 CAD / Case of 500
  • 5-9$603.02 CAD / Case of 500
  • 10+$569.52 CAD / Case of 500
$670.02CAD / Case of 500
BUY MORE & SAVE
  • 1-4$670.02 CAD / Case of 500
  • 5-9$603.02 CAD / Case of 500
  • 10+$569.52 CAD / Case of 500
Buy It Together
Current Item
DWK Life Sciences (Kimble) 73770-20125 Disposable Round Bottom Culture Tubes, 20 x 125, no markings, 500/cs
DWK Life Sciences (Kimble) 73770-20125 Disposable Round Bottom Culture Tubes, 20 x 125, no markings, 500/cs
3458058
$670.02CAD / Case of 500
+ =
Total Price 1111.30 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Tubes are offered with or without white markings
More About this Item
Manufactured of borosilicate glass, these disposable tubes are ideal for general bacteriological and tissue culture applications. Caps are sold separately.
Specifications
  • Capacity (mL)25
  • MaterialGlass
  • Cap / Stopper Size18-415
  • Label TypeNone
  • AutoclavableYes
  • Outside Diameter (OD) (cm)2
  • Overall Height (cm)12.5
  • DescriptionDisposable Round Bottom Culture Tubes, 20 x 125, no markings, 500/cs