Drager CMS Analyzer Chlorine, 0.2 to 10 ppm
Representative image only.
Drager CMS Analyzer Chlorine, 0.2 to 10 ppm
Drager | Mfr: 6406010| Item: 8651322
Eliminate user error for a consistent measurement every time
Call For Price
Call For Price
Buy It Together
Current Item
Drager CMS Analyzer Chlorine, 0.2 to 10 ppm
Drager CMS Analyzer Chlorine, 0.2 to 10 ppm
8651322
Call For Price
+ =
Total Price 0 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Digital display of gas concentration eliminates ambiguous results
  • Internal recorder stores up to 50 measurements with time/date stamp
  • 2 year warranty
More About this Item
Just load a CMS gas specific chip into the bottom port of the analyzer. Easy-to-use, three-step slide switch allows analyzer to take measurement. Gas concentration is displayed in seconds with digital readout. Operates on four AA batteries (included). Chips contain 10 tests each and are sold separately.
Two year warranty
Specifications
  • Gas MeasuredChlorine
  • Range0.2 to 10 ppm
  • DescriptionChlorine, 0.2 to 10 ppm