Digi-Sense Type K Economic Bolt On Thermocouple Probe #6 Screw with Mini-Connector, 36" Fiberglass Ext. Grounded
Representative image only.
Digi-Sense Type K Economic Bolt On Thermocouple Probe #6 Screw with Mini-Connector, 36" Fiberglass Ext. Grounded
Digi-Sense | Item: 1852638
Economical Bolt-On probes designed to be an easy and cost effective way to mount on a surface
$68.81CAD / Each
BUY MORE & SAVE
  • 1-1$68.81 CAD / Each
  • 2-9$61.93 CAD / Each
  • 10+$58.49 CAD / Each
for an additional
$340.35CAD / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
$68.81CAD / Each
BUY MORE & SAVE
  • 1-1$68.81 CAD / Each
  • 2-9$61.93 CAD / Each
  • 10+$58.49 CAD / Each
Buy It Together
Current Item
Digi-Sense Type K Economic Bolt On Thermocouple Probe #6 Screw with Mini-Connector, 36" Fiberglass Ext. Grounded
Digi-Sense Type K Economic Bolt On Thermocouple Probe #6 Screw with Mini-Connector, 36" Fiberglass Ext. Grounded
1852638
$68.81CAD / Each
+ =
Total Price 532.93 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Available in #6 (M3.5) or #8 (M4) screw sizes and different lead lenghts
  • Response Time: 2-4 seconds or greater
  • Each probe comes with a color coded ANSI mini connector or stripped leads: type J-black, type K-yellow, type T-blue, and type E-purple.
  • Custom designs or changes to this product are available upon request
  • 6 month warranty
More About this Item
Note: the temperature range is for the probe tip only. The handle material may be affected if the probe is rated higher than the handle depending on exposure time, application, and handle material. ABS handles are rated to 220°F (104°C); PVC handles are rated to 200°F (93°C); Stainless Steel handles are rated at 450°F (232°C); Mini Connectors are rated to 392°F (200°C).
6 Month Warranty
Specifications
  • Probe TypeK
  • Min Temperature (° F)-250
  • Max Temperature (° F)900
  • Min Temperature (° C)-150
  • Max Temperature (° C)482
  • Temperature Accuracy±3.0°F (1.1°C) or ±0.4% reading above 212°F(100°C); ±3.6°F (2.2°C) or 2% of reading between -58 to 211°F (-50 to 99°C); ±7.2°F (4°C) or ±4% of reading between -60 to -238 (-51 to -150°C), whichever is greater
  • JunctionGrounded
  • Connector TypeMini Connector
  • Sheath Material24 AWG, Fiberglass Insulated
  • DescriptionType K Econ Bolt On Thermocouple Probe Probe #6 Scr with Mini Con, 36" Fiberglass Ext. Grn