Digi-Sense Type-K Bolt-On Probe Mini-Connector, Grounded 5ft Strt 304 SS Armoredor/Fib Cable
Representative image only.
Digi-Sense Type-K Bolt-On Probe Mini-Connector, Grounded 5ft Strt 304 SS Armoredor/Fib Cable
Digi-Sense | Item: 0851685
Heavy Duty Bolt-On Probes for Long Term Installation
$104.32CAD / Each
BUY MORE & SAVE
  • 1-5$104.32 CAD / Each
  • 6-11$93.89 CAD / Each
  • 12-23$88.67 CAD / Each
  • 24+$83.46 CAD / Each
for an additional
$340.35CAD / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
$104.32CAD / Each
BUY MORE & SAVE
  • 1-5$104.32 CAD / Each
  • 6-11$93.89 CAD / Each
  • 12-23$88.67 CAD / Each
  • 24+$83.46 CAD / Each
Buy It Together
Current Item
Digi-Sense Type-K Bolt-On Probe Mini-Connector, Grounded 5ft Strt 304 SS Armoredor/Fib Cable
Digi-Sense Type-K Bolt-On Probe Mini-Connector, Grounded 5ft Strt 304 SS Armoredor/Fib Cable
0851685
$104.32CAD / Each
+ =
Total Price 568.44 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Each Probe comes with a bolt on washer that is .62” OD and .28” ID for quick easy installation
  • Once installed, the rugged design allows for continuous long term readings
  • This probe is not recommended for damp or wet conditions.
  • Response Time: 3 seconds or greater
  • Each probe comes with a color coded ANSI mini connector: type J-black, type K-yellow, type T-blue, and type E-purple.
  • Custom designs or changes to this product are available upon request.
  • 6 month warranty
More About this Item
Note: the temperature range is for the probe tip only. The handle material may be affected if the probe is rated higher than the handle depending on exposure time, application, and handle material. ABS handles are rated to 220°F (104°C); PVC handles are rated to 200°F (93°C); Stainless Steel handles are rated at 450°F (232°C); Mini Connectors are rated to 392°F (200°C).
6 Month Warranty
Specifications
  • Probe TypeK
  • Min Temperature (° F)-250
  • Max Temperature (° F)900
  • Min Temperature (° C)-150
  • Max Temperature (° C)482
  • Temperature Accuracy±3.0°F (1.1°C) or ±0.4% reading above 212°F(100°C); ±3.6°F (2.2°C) or 2% of reading between -58 to 211°F (-50 to 99°C); ±7.2°F (4°C) or ±4% of reading between -60 to -238 (-51 to -150°C), whichever is greater
  • JunctionGrounded
  • Connector TypeMini Connector
  • Sheath Material304 Stainless Steel Washer
  • Cable TypeSS armored fiberglass
  • Cable Length (InchShort)60
  • DescriptionType-K Hd Bolt-On Probe Mini Conn Grd 5ft Strt 304 SS Armoredor/Fib Cable