Digi-Sense Thermocouple Connector, Standard, Type-J, Female Jack; 1/ea
Representative image only.
Digi-Sense Thermocouple Connector, Standard, Type-J, Female Jack; 1/ea
Digi-Sense | Item: 1852709
Jacks and plugs good to 300°F (150°C)
$18.86CAD / Each
BUY MORE & SAVE
  • 1-1$18.86 CAD / Each
  • 2-9$16.97 CAD / Each
  • 10+$16.03 CAD / Each
$18.86CAD / Each
BUY MORE & SAVE
  • 1-1$18.86 CAD / Each
  • 2-9$16.97 CAD / Each
  • 10+$16.03 CAD / Each
Buy It Together
Current Item
Digi-Sense Thermocouple Connector, Standard, Type-J, Female Jack; 1/ea
Digi-Sense Thermocouple Connector, Standard, Type-J, Female Jack; 1/ea
1852709
$18.86CAD / Each
+
Traceable Monitoring Hygrometer
2025030
$91.20CAD / Each
=
Total Price 110.06 ADD 2 ITEMS TO CART
Customers Also Bought
Product Description
Key Features
  • Glass-filled nylon construction with molded barriers to prevent short circuit
  • Secure thermocouple wire into positive/negative terminals with screws
  • Each connector comes in ANSI color coded material: type J-black, type K-yellow, and type T-blue
  • 6 month warranty
More About this Item
Each connector comes in ANSI color coded material: type J-black, type K-yellow, and type T-blue.
6 Month Warranty
Specifications
  • Connector TypeJ
  • Special InformationGlass filled Nylon body
  • Max Temperature (° F)300
  • Max Temperature (° C)149
  • DescriptionThermocouple Connector, Standard, Type-J, Female Jack; 1/ea