Digi-Sense Solar-Powered Adjustable-Angle Fahrenheit Thermometer; 2.5" L
Representative image only.
Digi-Sense Solar-Powered Adjustable-Angle Fahrenheit Thermometer; 2.5" L
Digi-Sense | Item: 9013002
$503.16CAD / Each
for an additional
$332.62CAD / Each
for an additional
$461.17CAD / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
$503.16CAD / Each
Buy It Together
Current Item
Digi-Sense Solar-Powered Adjustable-Angle Fahrenheit Thermometer; 2.5" L
Digi-Sense Solar-Powered Adjustable-Angle Fahrenheit Thermometer; 2.5" L
9013002
$503.16CAD / Each
+ =
Total Price 835.78 ADD 2 ITEMS TO CART
Product Description
Key Features
  • 1 year warranty
More About this Item
View readings from almost any angle by simply adjusting the angle of the head. Three-inch diameter adjustable head has an easy-to-read LCD. Requiring no batteries these thermometers are ideal in pilot plant or research and development applications where exact readings are required. The safety circuit prevents false readings.
Each thermometer is housed in a weather-resistant 304 stainless steel (SS) case. Stem is 304 SS. An acrylic lens protects the face. To further protect thermometer order a thermowell from page 1052. Simply match the stem length to the probe depth "A" for the thermowell.
NSF international certified1 Year Warranty
Specifications
  • Min Temperature (° F)-50
  • Max Temperature (° F)300
  • Division (° F)0.1
  • Dial / Case Material304 Stainless Steel
  • Stem Material304 Stainless Steel
  • DescriptionSolar-Powered Adjustable-Angle Fahrenheit Thermometer; 2.5" L