Digi-Sense Rolling Wheel Surface Thermocouple Probe, Exposed; Type K
Representative image only.
Digi-Sense Rolling Wheel Surface Thermocouple Probe, Exposed; Type K
Digi-Sense | Item: 0844531
Roller Probes Designed to Monitor a Wide Range of Flat Stationary or Moving Surfaces
$581.25CAD / Each
BUY MORE & SAVE
  • 1-5$581.25 CAD / Each
  • 6+$523.13 CAD / Each
for an additional
$340.35CAD / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
$581.25CAD / Each
BUY MORE & SAVE
  • 1-5$581.25 CAD / Each
  • 6+$523.13 CAD / Each
Buy It Together
Current Item
Digi-Sense Rolling Wheel Surface Thermocouple Probe, Exposed; Type K
Digi-Sense Rolling Wheel Surface Thermocouple Probe, Exposed; Type K
0844531
$581.25CAD / Each
+ =
Total Price 1045.37 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Use to monitor hot plates, furnaces, molds, or any other flat surface
  • Ideal for rotary or other moving surfaces
  • Response Time: 2 seconds or greater
  • Each probe comes with a color coded ANSI coiled cables or connectors: type J-black, type K-yellow, type T-blue, and type E-purple.
  • Custom designs or changes to this product are available upon request
  • 6 month warranty
More About this Item
Note: the temperature range is for the probe tip only. The handle material may be affected if the probe is rated higher than the handle depending on exposure time, application, and handle material. ABS handles are rated to 220°F (104°C); PVC handles are rated to 200°F (93°C); Stainless Steel handles are rated at 450°F (232°C); Mini Connectors are rated to 392°F (200°C).
6 Month Warranty
Specifications
  • Probe TypeK
  • Length (")2 13/32
  • Min Temperature (° F)-250
  • Max Temperature (° F)450
  • Min Temperature (° C)-150
  • Max Temperature (° C)232
  • Temperature Accuracy±3.0°F (1.1°C) or ±0.4% reading above 212°F(100°C); ±3.6°F (2.2°C) or 2% of reading between -58 to 211°F (-50 to 99°C); ±7.2°F (4°C) or ±4% of reading between -60 to -238 (-51 to -150°C), whichever is greater
  • JunctionExposed
  • Sheath Material304 Stainless Steel with PTFE Guard
  • Handle MaterialPVC
  • Sheath Length (in)2.4062
  • DescriptionRolling Wheel Surface Thermocouple Probe, Exposed; Type K