Digi-Sense Miniature Type-K Thermocouple Female Connector, 2 Pin SKU 1852715
Representative image only.
Digi-Sense Miniature Type-K Thermocouple Female Connector, 2 Pin SKU 1852715
Digi-Sense | Item: 1852715
Handles extreme environments up to 650°F (343°C)!
$23.72CAD / Each
BUY MORE & SAVE
  • 1-1$23.72 CAD / Each
  • 2-9$21.35 CAD / Each
  • 10+$20.16 CAD / Each
$23.72CAD / Each
BUY MORE & SAVE
  • 1-1$23.72 CAD / Each
  • 2-9$21.35 CAD / Each
  • 10+$20.16 CAD / Each
Product Description
Key Features
  • Molded barriers prevent short circuit and the solid pin design ensures good contact even in demanding applications
  • Miniature connectors are suited for small wires and mineral insulated cables
  • Connector-body color is brown, calibration type (J or K) is printed on the body
  • 6 month warranty
More About this Item
High temperature connector-body color is brown, calibration indication printed on body.
6 Month Warranty
Specifications
  • Connector TypeK
  • Special InformationGlass filled thermosetting body
  • Max Temperature (° F)650
  • Max Temperature (° C)343
  • DescriptionMiniature Type-K Thermocouple Female Connector, 2 Pin