Digi-Sense High Temperature Thermocouple Probe, Fiberglass Insulated Wire, 20G, Exposed, Stripped Leads, Type K; 120" L
Representative image only.
Digi-Sense High Temperature Thermocouple Probe, Fiberglass Insulated Wire, 20G, Exposed, Stripped Leads, Type K; 120" L
Digi-Sense | Item: 0811331
Flexible Insulated Thermocouple Probes with Stripped Wire Leads for a Wide Range of Temperature Monitoring
$73.96CAD / Each
BUY MORE & SAVE
  • 1-1$73.96 CAD / Each
  • 2-9$66.56 CAD / Each
  • 10+$62.87 CAD / Each
for an additional
$340.35CAD / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
$73.96CAD / Each
BUY MORE & SAVE
  • 1-1$73.96 CAD / Each
  • 2-9$66.56 CAD / Each
  • 10+$62.87 CAD / Each
Buy It Together
Current Item
Digi-Sense High Temperature Thermocouple Probe, Fiberglass Insulated Wire, 20G, Exposed, Stripped Leads, Type K; 120" L
Digi-Sense High Temperature Thermocouple Probe, Fiberglass Insulated Wire, 20G, Exposed, Stripped Leads, Type K; 120" L
0811331
$73.96CAD / Each
+ =
Total Price 538.08 ADD 2 ITEMS TO CART
Product Description
Key Features
  • Response Time: 1 seconds or greater
  • All insulated-wire probes include straight insulated cable with no handle and stripped leads.
  • Custom designs or changes to this product are available upon request
  • 6 month warranty
More About this Item
Fiberglass wire is not recommended for use in moist or wet environments.
Exposed-junction probes are not recommended for use with aqueous solutions.
Do not subject type J exposed-junction probes to moisture as they will rust
6 Month Warranty
Specifications
  • Length (")120
  • Probe TypeK
  • Min Temperature (° F)-250
  • Max Temperature (° F)1300
  • Min Temperature (° C)-150
  • Max Temperature (° C)704
  • Temperature Accuracy±3.0°F (1.1°C) or ±0.4% reading above 212°F(100°C); ±3.6°F (2.2°C) or 2% of reading between -58 to 211°F (-50 to 99°C); ±7.2°F (4°C) or ±4% of reading between -60 to -238 (-51 to -150°C), whichever is greater
  • JunctionExposed
  • Connector TypeStripped Leads
  • Sheath MaterialHi-Temp Fiberglass Insulated Wire
  • Sheath Length (in)120
  • Sheath Diameter (InchShort)0.032
  • Cable Wire Gauge (Ga)20
  • DescriptionHi-Temperature Thermocouple Probe, Fib-Ins, 20G, Exp, Stripped; Type K