Digi-Sense Compression Fitting; Probe Diameter 1/8"; 316 Stainless Steel; 1/8"NPT (M)
Representative image only.
Digi-Sense Compression Fitting; Probe Diameter 1/8"; 316 Stainless Steel; 1/8"NPT (M)
Digi-Sense | Item: 1852790
Fittings let you adjust the depth your probe extends into a media you are measuring
$32.56CAD / Each
BUY MORE & SAVE
  • 1-1$32.56 CAD / Each
  • 2-9$29.30 CAD / Each
  • 10+$27.68 CAD / Each
$32.56CAD / Each
BUY MORE & SAVE
  • 1-1$32.56 CAD / Each
  • 2-9$29.30 CAD / Each
  • 10+$27.68 CAD / Each
Product Description
Key Features
  • Available in both 316 Stainless Steel and Brass
  • 6 month warranty
More About this Item
Use fittings for straight-shaft probes and metal tubing only. Tighten compression nut to secure and completely seal probe. Replacement ferrules let you reuse a compression fitting when replacing a worn-out probe. Simply disassemble fitting, discard probe and old ferrule, install replacement ferrule over probe sheath, and retighten fitting. Connectors come in both 316 Stainless Steel and Brass.
6 Month Warranty
Specifications
  • Connector TypeCompression Fitting
  • Special Information316 Stainless Steel
  • Max Temperature (° F)1200
  • Max Temperature (° C)649
  • DescriptionCompression Fitting; Probe Diameter 1/8"; 316 Stainless Steel; 1/8"NPT (M)