Delmhorst F-6/6-30/1235 F-6/6-30 Analog Hay Moisture Meter, Range 6% - 30% with 10" Probe

Representative image only.

Delmhorst F-6/6-30/1235 F-6/6-30 Analog Hay Moisture Meter, Range 6% - 30% with 10" Probe

Delmhorst | Mfr: F-6/6-30/1235| Item: 5982314

Prevent hay spoilage and fire hazards

$630.61CAD / Each

$630.61CAD / Each

Buy It Together

Current Item

Delmhorst F-6/6-30/1235 F-6/6-30 Analog Hay Moisture Meter, Range 6% - 30% with 10" Probe
Delmhorst F-6/6-30/1235 F-6/6-30 Analog Hay Moisture Meter, Range 6% - 30% with 10" Probe
5982314

$630.61CAD / Each
+
Delmhorst 1235 10" Bale Prod
5982319

$119.49CAD / Each
+ =
Total Price 871.36 ADD 3 ITEMS TO CART

Customers Also Bought

Product Description

Includes

one 9 V battery.

F-6/6-30 meter plus 10" (25.4 cm) direct-connect bale prod (1235)

Key Features

  • Color-coded range at optimum moisture content shows at a glance if hay is safe to bale
  • Simplified controls are user friendly for entry-level use
  • Built-in calibration check ensures accurate readings
  • 1 year warranty

More About this Item

This hay moisture meter is ideal for agricultural applications where you need to know if your crop is ready for baling and safe storage. Color-coding the optimum range of 18 to 22% MC (moisture content) lets you know at a glance if you need to let the hay dry further or if it needs to be wetted before baling.
Available in two ranges to suit requirements for both US domestic (13 to 40% MC) and export (6 to 30% MC) markets.

1 Year Warranty
Specifications
  • Measurement TypePin
  • Moisture Range Non-Wood Scale (Min) (%)6
  • Moisture Range Non-Wood Scale (Max) (%)30
  • Display TypeAnalog
  • Battery1 9V (included)
  • Height (in)3
  • Width (in)8
  • Depth (in)1 5/8
  • DescriptionF-6/6-30 Analog Hay Moisture Meter, Range 6% - 30% with 10" Probe
  • Warranty1 year
Reviews
Q&A