Davis Instruments Extra Solar Power Kit
Representative image only.
Davis Instruments Extra Solar Power Kit
Davis Instruments | Mfr: 6616| Item: 6800028
Power and protect outdoor installations of Vantage weather consoles or Weather Envoy data loggers
$884.03CAD / Each
$884.03CAD / Each
Product Description
Key Features
  • 1 year warranty
More About this Item
Shelters are ideal for protecting and in some cases powering your Vantage Vue™ and Vantage Pro2™ weather stations, and Weather Envoy and Envoy8X™ instruments when installed outdoors.
Extra Solar Panel Kit 6616 provides additional charging power to the battery in the 68000-24 or 68000-26 kits for sites where sunlight is limited. The 5 watt solar panel is mounted on a weather-resistant shelter in which a wireless console or Weather Envoy can be mounted. Dimensions are 13-3/4" x 10" x 4-1/18" (349 x 254 x 106 mm).
What's included: 5-watt solar panel, weather-resistant shelter, mounting hardware, and 4-ft (1-m) cable.
Available for sale in the United States and Canada only.
1 Year Warranty
Specifications
  • DescriptionExtra Solar Power Kit