Davis Instruments 6490 UV Sensor for Vanatage Pro2 (wireless)
Representative image only.
Davis Instruments 6490 UV Sensor for Vanatage Pro2 (wireless)
Davis Instruments | Mfr: 6490| Item: 3224547
Add-on sensors provide accurate weather montioring
$777.43CAD / Each
$777.43CAD / Each
Product Description
Includes
Built-in level and 2-ft (0.6-m) cable.
Key Features
  • Detects UV radiation at wavelengths of 290 to 390 nm
  • Displays UV index, dose rate, and daily and accumulated dose
More About this Item
Get accurate, reliable weather monitoring from these quality add-on sensors.
The sensor measures the sunburning portion of the UV spectrum. Multi-layer filter provides a spectral response that closely matches the Erythema Action Spectrum. Diffuser provides excellent cosine response. Two-piece housing minimizes radiation heating, allows convection cooling of the sensor, and prevents the trapping of water or dust.
Compatible with Vantage Pro, Vantage Pro2 and Vantage Vue (already included with Vantage Pro Plus and Vantage Pro2 Plus).
Available for sale in the United States and Canada only.
Specifications
  • DescriptionUV Sensor for Vanatage Pro2 (wireless)