CSC Scientific Dual Direction Tensiometer, ±0.1 dyn/cm
Representative image only.
CSC Scientific Dual Direction Tensiometer, ±0.1 dyn/cm
CSC Scientific | Mfr: 70545000| Item: 5995125
Ideal for measuring the surface tension of oils, detergents, colloidal suspensions, and chemicals
$14,431.86CAD / Each
for an additional
$330.41CAD / Each
for an additional
$453.80CAD / Each
Time to Re-Calibrate Your Instrument?
Learn more about our calibration services
$14,431.86CAD / Each
Buy It Together
Current Item
CSC Scientific Dual Direction Tensiometer, ±0.1 dyn/cm
CSC Scientific Dual Direction Tensiometer, ±0.1 dyn/cm
5995125
$14,431.86CAD / Each
+
InnoCal NIST-Traceable Calibration; Penetrometers
1705032
$330.41CAD / Each
+ + + =
Total Price 46189.67 ADD 5 ITEMS TO CART
Customers Also Bought
Product Description
Includes
a 6-cm platinum-iridium measuring ring, three extra torsion wires, and a carrying case.
Key Features
  • Measure surface tension in 15 to 30 seconds
  • Direct-reading scale in dyne/cm
  • 1 year warranty
More About this Item
Measure both the surface and interfacial tension of liquids. Range is 0 to 90 dyn/cm. Measurement values are reproducible to within 0.05 dyn/cm. These meters are specified for determining interfacial tension of oil against water by the ring method in ASTM D971, solutions of surface-active agents in ASTM D1331, testing synthetic rubber lattices in ASTM D1417, and determining surface tension of industrial water/wastewater in ASTM D1590. Model 59951-25 measures both upward and downward forces on the measuring ring—great for testing two-phase liquids. Features graduated dial readings in both directions from 0 to 90 dyn/cm and a double vernier. Both the scales and the corresponding verniers have graduations and figures in different colors to indicate whether the force is in an upward or downward direction.
1 Year Warranty
Specifications
  • Accuracy±0.1 dyn/cm
  • Range0 to 90 dyn/cm
  • DescriptionDual Direction Tensiometer, ±0.1 dyn/cm