Cryogenic label for laser printers, 1 9/32"W x 1/2"H
Representative image only.
Cryogenic label for laser printers, 1 9/32"W x 1/2"H
| Item: 6220670
$223.59CAD / Pkg of 1700
BUY MORE & SAVE
  • 1-5$223.59 CAD / Pkg of 1700
  • 6-29$201.23 CAD / Pkg of 1700
  • 30-119$190.05 CAD / Pkg of 1700
  • 120+$178.87 CAD / Pkg of 1700
$223.59CAD / Pkg of 1700
BUY MORE & SAVE
  • 1-5$223.59 CAD / Pkg of 1700
  • 6-29$201.23 CAD / Pkg of 1700
  • 30-119$190.05 CAD / Pkg of 1700
  • 120+$178.87 CAD / Pkg of 1700
Product Description
Key Features
  • 1 year warranty
More About this Item
The heat resistant, stay-flat sheets will not jam in laser printers
1 Year Warranty
Specifications
  • Temperature Points (° C)-196 to 150
  • Initial colorWhite
  • Width (in)1/2
  • Length (in)1 9/32
  • DescriptionCryogenic label for laser printers, 1 9/32"W x 1/2"H