Cryogenic label for laser printers, 1 11/16"W x 3/4"H
Representative image only.
Cryogenic label for laser printers, 1 11/16"W x 3/4"H
| Item: 6220672
$226.90CAD / Pkg of 1040
BUY MORE & SAVE
  • 1-5$226.90 CAD / Pkg of 1040
  • 6-29$204.21 CAD / Pkg of 1040
  • 30-119$192.87 CAD / Pkg of 1040
  • 120+$181.52 CAD / Pkg of 1040
$226.90CAD / Pkg of 1040
BUY MORE & SAVE
  • 1-5$226.90 CAD / Pkg of 1040
  • 6-29$204.21 CAD / Pkg of 1040
  • 30-119$192.87 CAD / Pkg of 1040
  • 120+$181.52 CAD / Pkg of 1040
Product Description
Key Features
  • 1 year warranty
More About this Item
The heat resistant, stay-flat sheets will not jam in laser printers
1 Year Warranty
Specifications
  • Temperature Points (° C)-196 to 150
  • Initial colorWhite
  • Height (in)3/4
  • Width (in)3/4
  • Length (in)1 11/16
  • DescriptionCryogenic label for laser printers, 1 11/16"W x 3/4"H