CPC (Colder) MicroCNX® Quick-Disconnect Fitting, Micro-Flow, Genderless Hose Barb, Polycarbonate, 1/8" ID

Representative image only.

CPC (Colder) MicroCNX® Quick-Disconnect Fitting, Micro-Flow, Genderless Hose Barb, Polycarbonate, 1/8" ID

CPC (Colder) | Mfr: CNX17002HT| Item: 5011215

Micro-flow genderless sterile connectors simplify process integration while maximizing flexibility

$55.84CAD / Each

$55.84CAD / Each

Buy It Together

Current Item

CPC (Colder) MicroCNX® Quick-Disconnect Fitting, Micro-Flow, Genderless Hose Barb, Polycarbonate, 1/8" ID
CPC (Colder) MicroCNX® Quick-Disconnect Fitting, Micro-Flow, Genderless Hose Barb, Polycarbonate, 1/8" ID
5011215

$55.84CAD / Each
+ + =
Total Price 145.57 ADD 3 ITEMS TO CART
Product Description

Key Features

  • Simple “pinch, click, and pull” design minimizes operator errors
  • Simplified tubing connection provides high performance and reliability
  • Designed for micro-flow media transfer applications
  • Free technical application support available to determine which connector type is needed for your application
  • Ideal for general-purpose, laboratory, and single-use applications
  • 1/8" hose barb

More About this Item

The MicroCNX® genderless connectors provide fast, easy, secure connections and quick assembly. The interior of the flow path is never exposed to the exterior environment providing sterile connections, even in non-sterile environments. Connectors eliminate the need for tube welding, saving you time and money. To use, simply pinch to remove the protective cover, join two halves together and squeeze until you hear an audible “click”, then pull the membrane strips away from the connector.

These aseptic quick connectors are made from animal-derived component-free materials, making them ideal for a wide range of biopharmaceutical, bioprocessing, cell therapy, gene therapy, and other critical-use applications.

Warning: This product is not approved or intended for, and should not be used for, medical, clinical, surgical or other patient-oriented applications.

Certificate of Compliance
Specifications
  • Fitting MaterialPolycarbonate
  • Seal MaterialPlatinum-Cured Silicone
  • Sterilization MethodGamma irradiation up to 50 kGy or autoclave one cycle up to 130°C (266°F) for 60 minutes
  • Connection TypeQuick Disconnect x Hose Barb
  • Barb Size / Tubing Id (Port 1) (mm)3.2
  • Barb Size / Tubing Id (Port 1) (in)1/8
  • Flow PatternStraight
  • ColorWhite with teal protective cover
  • Min Temperature (° F)34
  • Max Temperature (° F)104
  • Min Temperature (° C)1
  • Max Temperature (° C)40
  • Max Operating Pressure (PSI)60
  • Fitting Length (in)2 1/4
  • Fitting Width (in)1 31/64
  • Fitting Depth (in)59/64
  • Fitting Length (mm)57.2
  • Fitting Width (mm)37.8
  • Fitting Depth (mm)23.6
  • Regulatory ApprovalsFDA;RoHS;USP;ADCF;Non-DEHP;Non-Phthalate
  • DescriptionQuick-Disconnect Fitting, Micro-Flow, Genderless Hose Barb, Polycarbonate, 1/8" ID
  • WarrantyManufacturer defect
Documents & Certificates
Reviews
Q&A