Corning Universal Pipette Tips, 1 to 200 µL, Yellow, Non-Sterile; 1000/Pk
Representative image only.
Corning Universal Pipette Tips, 1 to 200 µL, Yellow, Non-Sterile; 1000/Pk
Corning | Mfr: 4866| Item: 2485082
Reliable fit with all major brand pipettors
$86.19CAD / Pkg of 1000
$86.19CAD / Pkg of 1000
Product Description
Key Features
  • Beveled orifice helps ensure accurate fluid delivery
  • Autoclavable
More About this Item
Corning® universal fit bulk packed polypropylene pipet tips are designed to assure a reliable fit with all major brand pipettors. Graduated at the 10, 50 and 100 µL volumes for quick and easy measurment readings.
Specifications
  • SterileNo
  • AutoclavableYes
  • Max Volume (µL)200
  • Min Volume (µL)1
  • Graduation Interval10, 50 and 100 µL
  • Tip StyleUniversal
  • Wetted MatterialPolypropylene
  • Color CodeYellow
  • Filter TypeNone
  • DescriptionUniversal Pipette Tips, 1 to 200 µL, Yellow, Non-Sterile; 1000/Pk