Corning Pyrex 1395-45LTC Polypropylene Caps for 1395 Media Bottles, Orange
Representative image only.
Corning Pyrex 1395-45LTC Polypropylene Caps for 1395 Media Bottles, Orange
DWK Life Sciences (Kimble) | Mfr: 1395-45LTC| Item: 3451450
Find replacements or upgrade your bottle's caps and pouring rings
$156.18CAD / Case of 20
BUY MORE & SAVE
  • 1-4$156.18 CAD / Case of 20
  • 5-9$140.56 CAD / Case of 20
  • 10+$124.94 CAD / Case of 20
$156.18CAD / Case of 20
BUY MORE & SAVE
  • 1-4$156.18 CAD / Case of 20
  • 5-9$140.56 CAD / Case of 20
  • 10+$124.94 CAD / Case of 20
Product Description
Key Features
  • Ensure the bottle cap size is the same as the bottle neck size
  • Polypropylene is autoclavable, PBT and ETFE can withstand dry heat sterilization
Specifications
  • Cap MaterialPolypropylene (PP)
  • Cap / Stopper SizeGL45
  • Cap ColorOrange
  • AutoclavableYes
  • DescriptionPolypropylene Caps for Brand 1395 Media Bottles, orange