Corning Pyrex 1395-45HTC Polypropylene Caps for 1395 Media Bottles, High Temperature, Red
Representative image only.
Corning Pyrex 1395-45HTC Polypropylene Caps for 1395 Media Bottles, High Temperature, Red
DWK Life Sciences (Kimble) | Mfr: 1395-45HTC| Item: 3451455
Find replacements or upgrade your bottle's caps and pouring rings
$275.52CAD / Case of 10
BUY MORE & SAVE
  • 1-4$275.52 CAD / Case of 10
  • 5-9$247.97 CAD / Case of 10
  • 10+$220.42 CAD / Case of 10
$275.52CAD / Case of 10
BUY MORE & SAVE
  • 1-4$275.52 CAD / Case of 10
  • 5-9$247.97 CAD / Case of 10
  • 10+$220.42 CAD / Case of 10
Product Description
Key Features
  • Ensure the bottle cap size is the same as the bottle neck size
  • Polypropylene is autoclavable, PBT and ETFE can withstand dry heat sterilization
Specifications
  • Cap MaterialPolybutylene Terephthalate (PBT)
  • Cap / Stopper SizeGL45
  • Cap ColorRed
  • Withstands High Temperature (DegreeF)356
  • AutoclavableYes
  • DescriptionPolypropylene Caps for Brand 1395 Media Bottles, high temperature, red