Corning Gosselin Bi-Plate Petri Dish, Polystyrene, 100 x 15 mm, Sterile; 825/Cs

Representative image only.

Corning Gosselin Bi-Plate Petri Dish, Polystyrene, 100 x 15 mm, Sterile; 825/Cs

Corning Gosselin | Mfr: BI93D-102| Item: 3455205

Optimized for automation and double-bagged for security

$302.04CAD / Case of 825

$302.04CAD / Case of 825
Product Description

Key Features

  • Optimal flatness ensures constant depth of agar
  • Three vents for easy gas exchange
  • Class 100 manufacturing environment ensures aseptic production and packaging
  • Double outer bag packaged 33 dishes/bag, 825/case

More About this Item

Bi-plate petri dishes are designed for use with automatic filling equipment. Dishes are constructed of crystal polystyrene with the injection point located on the side of the dish bottom to optimize clarity even by image analyzing systems.

Dishes are available in aseptic and sterile versions and feature cleanroom packaging with two outer bags and bar code label. Aseptic products are manufactured in a controlled environment, without human intervention during the manufacturing process. Sterile products are sterilized using an irradiation method compliant with the requirements of ANSI/AAMI/ISO 11137 to achieve (SAL 10⁻⁶).

Specifications
  • MaterialPolystyrene
  • ClosureVented
  • Height (mm)15
  • Diameter (mm)100
  • SterileSterile
  • DescriptionBi-Plate Petri Dish, Polystyrene, 100 x 15 mm, Sterile; 825/Cs
Reviews
Q&A