Corning 96-Well Microplate without Lid, Polystyrene, Non-Sterile, Clear, Not Treated, V-Bottom; 100/Cs

Representative image only.

Corning 96-Well Microplate without Lid, Polystyrene, Non-Sterile, Clear, Not Treated, V-Bottom; 100/Cs

Corning | Mfr: 3897| Item: 0172805

Surfaces with different degrees of hydrophilicity—ideal for solid phase immunoassays (ELISA)

$565.78CAD / Case of 100

$565.78CAD / Case of 100

Buy It Together

Current Item

Corning 96-Well Microplate without Lid, Polystyrene, Non-Sterile, Clear, Not Treated, V-Bottom; 100/Cs
Corning 96-Well Microplate without Lid, Polystyrene, Non-Sterile, Clear, Not Treated, V-Bottom; 100/Cs
0172805

$565.78CAD / Case of 100
+ =
Total Price 712.82 ADD 2 ITEMS TO CART
Product Description

Key Features

  • Formulated for uniform binding and low background absorption
  • Conforms to standard microplate footprint and dimensions
  • Constructed of durable polystyrene

More About this Item

These 96-well polystyrene microplates meet the SBS standard to ensure compatibility with microplate and automation equipment. Plates are available sterile and non-sterile, with and without lids, and come in a wide range of colors, surface treatments, and well shapes.

Plate Colors: Clear plates have excellent optical properties making it ideal for optical measurements. Black plates have low background fluorescence, minimal light scatter and reduced crosstalk. White plates enhance luminescent signals and have low background luminescence and fluorescence.

Surface Treatment options are the standard tissue culture (TC) surface treatment for optimal cell attachment, not treated/medium binding surface treatment which is hydrophobic in nature and binds biomolecules through passive interactions, and high binding treatment which is capable of binding medium (>10kD) and large biomolecules that possess ionic groups and/or hydrophobic regions.

Well Shape options include flat wells which maximize light transmission, round wells which optimize coating and washing, and V-bottom wells let you completely retrieve well contents.

Specifications
  • Number of Wells96
  • Well Volume (µL)320
  • Min Working Well Volume (mL)0.075
  • Max Working Well Volume (mL)0.2
  • Well Bottom Diameter (mm)6.35
  • Well Top Diameter (mm)6.9
  • Well ShapeV-Bottom
  • Surface TreatmentNot Treated
  • Lid (Included)No
  • SterileNo
  • Plate ColorClear
  • Binding PropertyMedium
  • MaterialPolystyrene
  • Height (in)9/16
  • Width (in)3 3/8
  • Length (in)5 1/32
  • Packaging25 Plates/Pack, 100 Plates/Case
  • Description96-Well Microplate without Lid, Polystyrene, Non-Sterile, Clear, Not Treated, V-Bottom; 100/Cs
Reviews
Q&A