Corning 4807 IsoTip, 0.1 to 10 µL, G tip, sterile, 960/pk
Representative image only.
Corning 4807 IsoTip, 0.1 to 10 µL, G tip, sterile, 960/pk
Corning | Mfr: 4807| Item: 2485090
Prevents cross-contamination in critical application
$218.06CAD / Pkg of 960
$218.06CAD / Pkg of 960
Product Description
Key Features
  • Packaged sterile
  • RNase-/DNase-free
  • Nonpyrogenic
More About this Item
These IsoTips feature an inert, hydrophobic barrier which prevents aerosolized contaminants from coming in contact with pipettor shafts,. They are manufactured using Statistical Process Control (SPC) in their modeling operation to ensure premium quality. This process yields consistent critical dimensions such as tip straightness, length, and sealing ring location. Tips will properly attach to and eject from your pipettor everytime you use them. Ideal for preventing cross contamination is critical, such as DNA amplification and radioisotope handling.
Specifications
  • SterileYes
  • Max Volume (µL)10
  • Min Volume (µL)0.1
  • Min Volume (mL)0.0001
  • Max Volume (mL)0.1
  • Tip Volume (mL)0.01
  • Min Dispensing (mL)0.0001
  • Max Dispensing (mL)0.01
  • DescriptionIsoTip, 0.1 to 10 µL, G tip, sterile, 960/pk