Compound vials, 14.75 x 22 mm, 1.25 mL, amber glass, 144/cs

Representative image only.

Compound vials, 14.75 x 22 mm, 1.25 mL, amber glass, 144/cs

| Mfr: GLC-08767| Item: 0375722

Ideal for storage of small amounts of compounds such as reference standards

$348.09CAD / Case of 144

$348.09CAD / Case of 144
Product Description

Key Features

  • Safely and securely store samples

More About this Item

These borosilicate glass vials are available in clear or amber glass. Vials are low profile for greater stability and wide mouth for ease of sampling. Cases contain a shrink-wrapped tray vials including caps.

Specifications
  • Volume (mL)1.25
  • Tube MaterialBorosilicate glass
  • Tube Bottom ShapeFlat
  • Tube ColorAmber
  • Height (in)7/8
  • Outside Diameter (in)37/64
  • Height (mm)22
  • Outside Diameter (mm)14.75
  • DescriptionCompound vials, 14.75 x 22 mm, 1.25 mL, amber glass, 144/cs
Reviews
Q&A